4,5-Dimethoxy-1H-indole-2-carboxylic acid 95%
Supplier: Apollo Scientific
Warning
OR13450-250MG OR13450-1G OR13450-5G
APOSOR13450-250MGEA 87 EUR
APOSOR13450-250MG APOSOR13450-1G APOSOR13450-5G
4,5-Dimethoxy-1H-indole-2-carboxylic acid 95%
4,5-Dimethoxy-1H-indole-2-carboxylic acid
Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
Melting Pt: 222…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02664472
CAS Number: 50536-49-7