trans,trans-2,4-Hexadien-1-ol 95%
Supplier: Apollo Scientific
Danger
Synonyms: (2E,4E)-Hexa-2,4-dien-1-ol, Trans,trans-2,4-hexadien-1-ol, (2E,4E)-1-Hydroxyhexa-2,4-diene, Sorbic alcohol, trans,trans-Hexa-2,4-dien-1-ol
OR13712-10G OR13712-5G
APOSOR13712-10GEA 42.2 EUR
APOSOR13712-10G APOSOR13712-5G
trans,trans-2,4-Hexadien-1-ol 95%
trans,trans-2,4-Hexadien-1-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 80/12 mm °C
Melting Pt: 27…33 °C
Density: 0.871
Flash Pt: 72 °C
MDL Number: MFCD00002925
CAS Number: 17102-64-6
UN: 2811
ADR: 6.1,III