3-(1,1-Difluoroethyl)phenacyl bromide 95%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-1-[3-(1,1-difluoroethyl)phenyl]ethanone
PC56998-250MG PC56998-1G PC56998-5G
APOSPC56998-250MGEA 174 EUR
APOSPC56998-250MG APOSPC56998-1G APOSPC56998-5G
3-(1,1-Difluoroethyl)phenacyl bromide 95%
3-(1,1-Difluoroethyl)phenacyl bromide
Formula: C₁₀H₉BrF₂O
MW: 263.08 g/mol
UN: 3265
ADR: 8 II