1-Fluoro-4-nitrobenzene 96%
Supplier: Apollo Scientific
Danger
PC57472-250G PC57472-1KG
APOSPC57472-250GEA 23.2 EUR
APOSPC57472-250G APOSPC57472-1KG
1-Fluoro-4-nitrobenzene 96%
Formula: C₆H₄FNO₂
Boiling Pt: 205 °C
Melting Pt: 21 °C
Density: 1.33
Flash Pt: 90 °C
MDL Number: MFCD00007282
CAS Number: 350-46-9
UN: 2811
ADR: 6.1,III