Methyl 2-bromo-5-fluoro-3-(trifluoromethyl)benzoate 95%
Supplier: Apollo Scientific
Warning
PC57800-250MG PC57800-1G PC57800-5G
APOSPC57800-250MGEA 154 EUR
APOSPC57800-250MG APOSPC57800-1G APOSPC57800-5G
Methyl 2-bromo-5-fluoro-3-(trifluoromethyl)benzoate 95%
Methyl 2-bromo-5-fluoro-3-(trifluoromethyl)benzoate
Formula: C₉H₅BrF₄O₂
MW: 301.03 g/mol
MDL Number: MFCD32903691
CAS Number: 2091662-04-1