4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-6-(tert-butyl)-3-mercapto-1,2,4-triazin-5(4H)-one
OR24684-250MG OR24684-100MG
APOSOR24684-250MGEA 24.8 EUR
APOSOR24684-250MG APOSOR24684-100MG
4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one 97%
4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one
Formula: C₇H₁₂N₄OS
MDL Number: MFCD00225752
CAS Number: 33509-43-2