4-Formyltetrahydropyran 90%
Supplier: Apollo Scientific
Warning
Synonyms: Tetrahydropyran-4-carbaldehyde, Tetrahydro-2H-pyran-4-carboxaldehyde
OR31055-1G OR31055-5G OR31055-25G
APOSOR31055-1GEA 21.9 EUR
APOSOR31055-1G APOSOR31055-5G APOSOR31055-25G
4-Formyltetrahydropyran 90%
4-Formyltetrahydropyran
Formula: C₆H₁₀O₂
MW: 114.1424 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03425264
CAS Number: 50675-18-8