2-(Cyclopropyl)-5-methoxybenzoic acid 95%
Supplier: Apollo Scientific
Warning
OR315847-250MG OR315847-1G OR315847-5G
APOSOR315847-250MGEA 174 EUR
APOSOR315847-250MG APOSOR315847-1G APOSOR315847-5G
2-(Cyclopropyl)-5-methoxybenzoic acid 95%
2-(Cyclopropyl)-5-methoxybenzoic acid
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD16778842
CAS Number: 1250297-02-9