5-Cyclopropylpyridine-2-carboxaldehyde 95%
Supplier: Apollo Scientific
Warning
OR33673-250MG OR33673-1G
APOSOR33673-250MGEA 203 EUR
APOSOR33673-250MG APOSOR33673-1G
5-Cyclopropylpyridine-2-carboxaldehyde 95%
5-Cyclopropylpyridine-2-carboxaldehyde
Formula: C₉H₉NO
MW: 147.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18254230
CAS Number: 1256825-21-4