4-Acetylphenylboronic acid pinacol ester 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 4-Acetylphenylboronic acid, pinacol ester, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
OR360618-1G OR360618-5G OR360618-25G
APOSOR360618-1GEA 26.1 EUR
APOSOR360618-1G APOSOR360618-5G APOSOR360618-25G
4-Acetylphenylboronic acid pinacol ester 95%
4-Acetylphenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₃
Boiling Pt: 356.6/760 mm °C
Melting Pt: 63…65 °C
Density: 1.04
Flash Pt: 169.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05863922
CAS Number: 171364-81-1