1-(4-Bromo-3-nitrophenyl)-1-propanone 98%
Supplier: Apollo Scientific
Warning
OR450140-1G OR450140-5G
APOSOR450140-1GEA 163 EUR
APOSOR450140-1G APOSOR450140-5G
1-(4-Bromo-3-nitrophenyl)-1-propanone 98%
1-(4-Bromo-3-nitrophenyl)-1-propanone
Formula: C₉H₈BrNO₃
MDL Number: MFCD00600442
CAS Number: 101860-83-7