Sodium-L(+)-glutamate monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate
MQ200
G5889-100G G5889-1KG G5889-500G
SIALG5889-100GEA 49.5 EUR
SIALG5889-100G SIALG5889-1KG SIALG5889-500G
Sodium-L(+)-glutamate monohydrate, Sigma-Aldrich®
Sodium-L(+)-glutamate monohydrate
Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
Melting Pt: 225…240 °C
Flash Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278