L(-)-Proline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P0380-100G P0380-10MG P0380-1KG P0380-5KG
SIALP0380-100GEA 166 EUR
SIALP0380-100G SIALP0380-10MG SIALP0380-1KG SIALP0380-5KG
L(-)-Proline, Sigma-Aldrich®
L(-)-Proline
Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871