L(-)-Proline, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
M-Clarity™ quality level = MQ300
W331902-100G-K
W331902-1KG-K
W331902-5KG-K
SIALW331902-100G-KEA
77.8
EUR
SIALW331902-100G-K
SIALW331902-1KG-K
SIALW331902-5KG-K
L(-)-Proline, Sigma-Aldrich®
L(-)-Proline
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 220…222 °C Density: 1.35 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064318 CAS Number: 147-85-3 EINECS: 205-702-2 Merck Index: 13,07871 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...