Hematoxylin

Supplier: Molekula

MOLE29628570EA 125 EUR
MOLE29628570
Hematoxylin
Hematoxylin
Formula: C₁₆H₁₄O₆
MW: 302,28 g/mol
Melting Pt: 291…293 °C
MDL Number: MFCD00078111
CAS Number: 517-28-2
Merck Index: 13,04654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR