2-Methylfuran 99% stabilised
Supplier: Thermo Scientific
Danger
126830010 126830050 126832500
ACRO126830010EA 114 EUR
ACRO126830010 ACRO126830050 ACRO126832500
2-Methylfuran 99% stabilised
2-Methylfuran
Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 63…66 °C (760 mmHg)
Melting Pt: –88.7 °C
Density: 0.91 g/cm³
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00003248
CAS Number: 534-22-5
EINECS: 208-594-5
UN: 2301
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4340 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.5 %
Stabilizer 0.01 % hydroquinone