4-Phenoxyaniline 97%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Amino-4-phenoxybenzol
130231000 130230050 130235000
ACRO130231000EA 62.6 EUR
ACRO130231000 ACRO130230050 ACRO130235000
4-Phenoxyaniline 97%
4-Phenoxyaniline
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 140 °C (3hPa)
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007862
CAS Number: 139-59-3
EINECS: 205-367-2
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Brown to green-brown Powder or flakes
Infrared spectrum Conforms
Melting point 82 to 86 °C
Titration with HClO4 ≥96.0 %