Triethyl orthoformate 98%
Supplier: Thermo Scientific
Warning
139660010 139660025 139661000 139660250
ACRO139660010EA 66.1 EUR
ACRO139660010 ACRO139660025 ACRO139661000 ACRO139660250
Triethyl orthoformate 98%
Triethyl orthoformate
Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 146 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.89 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
EINECS: 204-550-4
UN: 2524
ADR: 3,III
Merck Index: 12,07012
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3900 to 1.3920 (20°C, 589 nm)
GC ≥97.5 %