N-(4-Bromobutyl)phthalimide 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione
146290100 146290500
ACRO146290100EA 88.7 EUR
ACRO146290100 ACRO146290500
N-(4-Bromobutyl)phthalimide 98%
N-(4-Bromobutyl)phthalimide
Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00005905
CAS Number: 5394-18-3
EINECS: 226-401-2
Order Now
Specification Test Results
Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %