Isobutanol ≥99% ACS
Supplier: Spectrum Chemical
Danger
SPCMB1186-100MLEA 119 EUR
SPCMB1186-100ML SPCMB1186-1LT SPCMB1186-500MLGL
Isobutanol ≥99% ACS
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74,12 g/mol
Boiling Pt: 107,9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0,803
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://si.vwr.com/stibo/web/std.lang.all/12/85/2301285.jpg
Order Now
Specification Test Results
Assay 99% Min.