9-Decen-1-ol 99%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Decen-10-ol
152640050 152640250 152641000
ACRO152640050EA 35.9 EUR
ACRO152640050 ACRO152640250 ACRO152641000
9-Decen-1-ol 99%
9-Decen-1-ol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 234…238 °C (760 mmHg)
Melting Pt: –13 °C
Density: 0.845 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002992
CAS Number: 13019-22-2
EINECS: 235-878-6
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥92 % (main isomer)