2-Chlorobenzothiazole 99%
Supplier: Thermo Scientific
Warning
173490250 AC17349.1 173490050
ACRO173490250EA 125 EUR
ACRO173490250 MAYBAC17349.1 ACRO173490050
2-Chlorobenzothiazole 99%
2-Chlorobenzothiazole
Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: 21…24 °C
Density: 1.37 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0
Order Now
Specification Test Results
Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6367 to 1.6387 (20°C, 589 nm)
GC ≥98.5 %