Tetrakis(triphenylphosphine)palladium(0) 99%
Supplier: Thermo Scientific
Warning
Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
202380010 202380050 202380250 202381000
202380010.EA 69.8 EUR
202380010. 202380050. 202380250. 202381000.
Tetrakis(triphenylphosphine)palladium(0) 99%
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Melting Pt: 103…107 °C (dec.)
Density: 1 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9
Order Now
Specification Test Results
Appearance Yellow to yellow-green Powder or crystals
Infrared spectrum Conforms
Carbon (C) 73.7 to 76.0 %