1-Phenyl-2-thiourea 97%
Supplier: Thermo Fisher Scientific
Danger
207250050 207250250 207251000
ACRO207250050EA 27.6 EUR
ACRO207250050 ACRO207250250 ACRO207251000
1-Phenyl-2-thiourea 97%
1-Phenyl-2-thiourea
Formula: C₇H₈N₂S
Boiling Pt: 295…297 °C (1013 hPa)
Melting Pt: 148…150 °C
Density: 1,33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004933
CAS Number: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,I
Merck Index: 12,07468
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %