Disodium succinate, anhydrous 99%
Supplier: Thermo Scientific
Warning
Synonyms: 1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic
208741000 208745000
ACRO208741000EA 28.4 EUR
ACRO208741000 ACRO208745000
Disodium succinate, anhydrous 99%
Disodium succinate
Formula: C₄H₄Na₂O₄
MW: 162.05 g/mol
Melting Pt: >300 °C
Density: 1,88 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002790
CAS Number: 150-90-3
EINECS: 205-778-7
Merck Index: 12,08828
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.3 % (120°C)
Titration with HClO4 ≥98.5 %