Tetrakis(triphenylphosphine)nickel(0) 95%
Supplier: Thermo Scientific
Danger
223980010 223980050
223980010.EA 120 EUR
223980010. 223980050.
Tetrakis(triphenylphosphine)nickel(0) 95%
Tetrakis(triphenylphosphine)nickel (0)
Formula: C₇₂H₆₀NiP₄
MW: 1107.85 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010011
CAS Number: 15133-82-1
UN: 1325
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Rust brown to brown Powder or crystals
Elemental Analysis ≥94 %