3,5-Bis(trifluoromethyl)benzylamine 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine
305640010 305640050
ACRO305640010EA 38.7 EUR
ACRO305640010 ACRO305640050
3,5-Bis(trifluoromethyl)benzylamine 95%
3,5-Bis(trifluoromethyl)benzylamine
Formula: C₉H₇F₆N
Boiling Pt: 82…84 °C (15 mmHg)
Melting Pt: 46…55 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009909
CAS Number: 85068-29-7
EINECS: 285-290-9
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
GC ≥94 %