4-Cyanophenyl isothiocyanate

Supplier: Thermo Scientific

Synonyms: 4-Cyanophenylisothiocyanate, 1-Cyano-4-isothiocyanatobenzene, 4-Isothiocyanatobenzonitrile

AC34897.1 AC34897.10
MAYBAC34897.1EA 0 EUR
MAYBAC34897.1 MAYBAC34897.10
4-Cyanophenyl isothiocyanate
4-Cyanophenyl isothiocyanate
Formula: C₈H₄N₂S
MW: 160.2 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 131…132 °C
MDL Number: MFCD00041085
CAS Number: 2719-32-6
EINECS: 220-323-2
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR