1-Ethyl-3-methylimidazolium bromide 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Ethyl-1-methyl-1H-imidazol-3-ium bromide
356670250 356670050 356671000
ACRO356670250EA 56.7 EUR
ACRO356670250 ACRO356670050 ACRO356671000
1-Ethyl-3-methylimidazolium bromide 98%
1-Ethyl-3-methylimidazolium bromide
Formula: C₆H₁₁BrN₂
MW: 191.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427610
CAS Number: 65039-08-9
Order Now
Specification Test Results
Appearance Yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %