Methylamine 2 mol/l in methanol, AcroSeal®, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Aminomethane
368551000 368558000
368551000.EA 130 EUR
368551000. 368558000.
Methylamine 2 mol/l in methanol, AcroSeal®, pure
Methylamine
Formula: CH₅N
Boiling Pt: 40 °C
Density: 0.785 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00008104
CAS Number: 74-89-5
EINECS: 200-820-0
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear solution
Strength 2 M (typically) exact strength on the certificate of analysis