3-Aminophthalimide

Supplier: Thermo Scientific

Synonyms: 4-Aminoisoindoline-1,3-dione

AC40100.1 AC40100.10
MAYBAC40100.1EA 0 EUR
MAYBAC40100.1 MAYBAC40100.10
3-Aminophthalimide
3-Aminophthalimide
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Boiling Pt: 447 °C (1013 hPa)
Melting Pt: 270…271 °C
MDL Number: MFCD00041853
CAS Number: 2518-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR