(±)-Tetrahydrofurfuryl alcohol 99+%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA
420440010 420442500 420440025
ACRO420440010EA 49.2 EUR
ACRO420440010 ACRO420442500 ACRO420440025
(±)-Tetrahydrofurfuryl alcohol 99+%
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 178 °C (760 mmHg)
Melting Pt: –80 °C
Density: 1.0543 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.0 %
Color scale ≤30 APHA