Diammonium tetrachloropalladate ≥99.998% (metals basis) Pd 37%, Premion®
Supplier: Thermo Scientific
Danger
Synonyms: Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)
10824.03 10824.06 10824.14
10824.03EA 138 EUR
10824.03 10824.06 10824.14
Diammonium tetrachloropalladate ≥99.998% (metals basis) Pd 37%, Premion®
Diammonium tetrachloropalladate
Purity: 99.998% CAS No.: 13820-40-1 Molecular Formula: H8Cl4N2Pd Formula Weight: 284.31 Physical Form: Powder Density: 2.17 Appearance: Dark green to brown Melting Point: dec. MDL No.: MFCD00010884
Formula: H₈Cl₄N₂Pd
MW: 284.31 g/mol
Melting Pt: Dec.
Density: 2.17
Storage Temperature: Ambient
MDL Number: MFCD00010884
CAS Number: 13820-40-1
EINECS: 237-498-6