Sodium hexachloropalladate(IV) hydrate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Disodium hexachloropalladate hydrate
11039.03 11039.06
11039.03EA 173 EUR
11039.03 11039.06
Sodium hexachloropalladate(IV) hydrate ≥99.9% (metals basis)
Disodium hexachloropalladate hydrate
Purity: 99.9% CAS No.: 53823-60-2 Molecular Formula: Cl6Na2Pd.xH2O Formula Weight: 365.10(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red to brown MDL No.: MFCD00014235
Formula: Cl₆Na₂Pd·xH₂O
MW: 365.10 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00014235
CAS Number: 53823-60-2
EINECS: 258-809-1