Formaldehyde 37% (w/w) in aqueous solution stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Stabilization:
Stabilised with methanol 7-8%
A16163.0F
A16163.AP
A16163.0FEA
57.3
EUR
A16163.0F
A16163.AP
Formaldehyde 37% (w/w) in aqueous solution stabilised
Formaldehyde
CAS No.: 50-00-0
Molecular Formula: CH2O
Formula Weight: 30.03
Flash Point: 60°(140°F)
Refractive Index: 1.3765
Odour: Pungent
Boiling Point: 97°
Beilstein Reference: 1209228
Merck Reference: 14,4235
UN No.: 1198
MDL No.: MFCD00003274
Formula:
CH₂O Boiling Pt: 97 °C Melting Pt: –15 °C Density: 1.083 Flash Pt: 66 °C (151 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 2209 ADR: 8,III Merck Index: 14,4235 |