2-Amino-6-fluorobenzothiazole 99%
Supplier: Thermo Scientific
Warning
Synonyms: 6-Fluorobenzothiazol-2-ylamine, 6-Fluorobenzo[d]thiazol-2-amine, 6-Fluoro-1,3-benzothiazol-2-amine
A18383.03 A18383.06 A18383.14
A18383.03EA 24.6 EUR
A18383.03 A18383.06 A18383.14
2-Amino-6-fluorobenzothiazole 99%
2-Amino-6-fluorobenzothiazole
Purity: 99% CAS No.: 348-40-3 Molecular Formula: C7H5FN2S Formula Weight: 168.19 Melting Point: 183-185° MDL No.: MFCD00013336
Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 183…185 °C
Density: 1.59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013336
CAS Number: 348-40-3