4-Ethoxyphenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-Ethoxy-4-isothiocyanato-benzene, 4-Ethoxyphenylisothiocyanate
A18902.06 A18902.14
A18902.06EA 80.1 EUR
A18902.06 A18902.14
4-Ethoxyphenyl isothiocyanate ≥98%
4-Ethoxyphenyl isothiocyanate
Purity: 98% CAS No.: 3460-49-9 Molecular Formula: C9H9NOS Formula Weight: 179.24 Flash Point: 110°(230°F) Odour: Irritating Beilstein Reference: 637526 UN No.: 2923 MDL No.: MFCD00041096
Formula: C₉H₉NOS
Boiling Pt: 146 °C (19 mmHg)
Melting Pt: 57…61 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041096
CAS Number: 3460-49-9
EINECS: 222-410-0
UN: 2923
ADR: 8,III