2-Benzyloxyphenol ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-(Benzyloxy)phenol, 2-Hydroxyphenyl benzyl ether
A19020.06 A19020.14
A19020.06EA 57.1 EUR
A19020.06 A19020.14
2-Benzyloxyphenol ≥98%
2-Benzyloxyphenol
Purity: 98% CAS No.: 6272-38-4 Molecular Formula: C13H12O2 Formula Weight: 200.24 Flash Point: 110°(230°F) Refractive Index: 1.5910 MDL No.: MFCD00002186
Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 336
Melting Pt: 38
Density: 1.143
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002186
CAS Number: 6272-38-4
EINECS: 228-461-5