Ursodeoxycholic acid ≥99%
Supplier: Thermo Fisher Scientific
Warning
B20490.03 B20490.06 B20490.14
B20490.03EA 42 EUR
B20490.03 B20490.06 B20490.14
Ursodeoxycholic acid ≥99%
Ursodeoxycholic acid
Purity: 99% CAS No.: 128-13-2 Molecular Formula: C24H40O4 Formula Weight: 392.56 Melting Point: 201-204° Merck Reference: 14,9889 MDL No.: MFCD00003680
Formula: C₂₄H₄₀O₄
Boiling Pt: 547 °C (1013 hPa)
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00003680
CAS Number: 128-13-2
EINECS: 204-879-3
Merck Index: 14,9889