8-Bromocaprylic acid ≥97%
Supplier: Thermo Fisher Scientific
B25575.09
B25575.18
B25575.09EA
133
EUR
B25575.09
B25575.18
8-Bromocaprylic acid ≥97%
8-Bromocaprylic acid
Purity: 97%
CAS No.: 17696-11-6
Molecular Formula: C8H15BrO2
Formula Weight: 223.11
Flash Point:
Learn more