2,4-Dimethoxypyrimidine ≥98%
Supplier: Thermo Scientific
|
Warning
|
H25799.06
H25799.03
H25799.06EA
95.2
EUR
H25799.06
H25799.03
2,4-Dimethoxypyrimidine ≥98%
2,4-Dimethoxypyrimidine
Purity: 98+%
CAS No.: 3551-55-1
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Flash Point: 93°(199°F)
Refractive Index: 1.502
Melting Point: 17°
Boiling Point: 204-205°
MDL No.: MFCD00056028
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol Boiling Pt: 204…205 °C Melting Pt: 17 °C Density: 1.1501 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00056028 CAS Number: 3551-55-1 |