2,6-Diisopropyl-N,N-dimethylaniline 96%
Supplier: Thermo Fisher Scientific
Danger
H27275.06 H27275.03
H27275.06EA 146 EUR
H27275.06 H27275.03
2,6-Diisopropyl-N,N-dimethylaniline 96%
2,6-Diisopropyl-N,N-dimethylaniline
Purity: 96% CAS No.: 2909-77-5 Molecular Formula: C14H23N Formula Weight: 205.34 Flash Point: 110°(230°F) Refractive Index: 1.5005 Boiling Point: 86°/2.5mm UN No.: 2810 MDL No.: MFCD00144978
Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Boiling Pt: 86 °C (2.5 mmHg)
Density: 0.873
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00144978
CAS Number: 2909-77-5
UN: 2810
ADR: 6.1 III