3-Bromo-p-toluonitrile ≥97%
Supplier: Thermo Fisher Scientific
Danger
H27806.14 H27806.06
H27806.14EA 145 EUR
H27806.14 H27806.06
3-Bromo-p-toluonitrile ≥97%
3-Bromo-p-toluonitrile
Purity: 97% CAS No.: 42872-74-2 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 110°(230°F) Melting Point: 39-43° UN No.: 3439 MDL No.: MFCD06797818
Formula: C₈H₆BrN
Melting Pt: 39…43 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06797818
CAS Number: 42872-74-2
UN: 3439
ADR: 6.1,III