1-(2-Pyridyl)piperazine ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
L05451.06
L05451.14
L05451.22
L05451.06EA
38.9
EUR
L05451.06
L05451.14
L05451.22
1-(2-Pyridyl)piperazine ≥99%
1-(2-Pyridyl)piperazine
Purity: 99%
CAS No.: 34803-66-2
Molecular Formula: C9H13N3
Formula Weight: 163.22
Flash Point:
Formula:
C₉H₁₃N₃ Boiling Pt: 120…122 °C (2 mmHg) Melting Pt: 85…87 °C Density: 1.072 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006216 CAS Number: 34803-66-2 EINECS: 252-220-3 ADR: 8,III |