2-Amino-5-metiltiazol ≥98%
Supplier: Thermo Scientific
|
Warning
|
L11039.06EA
26.6
EUR
L11039.06
2-Amino-5-metiltiazol ≥98%
2-Amino-5-metiltiazol
Formula:
C₄H₆N₂S MW: 114,17 g/mol Boiling Pt: 222 °C (1013 hPa) Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078317 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...