4,6-Dichloropyrimidine 98
Supplier: Thermo Fisher Scientific
Danger
L13212.06 L13212.14
L13212.06EA 59.2 EUR
L13212.06 L13212.14
4,6-Dichloropyrimidine 98
4,6-Dichloropyrimidine
Purity: 98% CAS No.: 1193-21-1 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Melting Point: 64-68° Boiling Point: 176° UN No.: 3261 MDL No.: MFCD00006109
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 176 °C
Melting Pt: 64…68 °C
Density: 1.734 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006109
CAS Number: 1193-21-1
EINECS: 214-770-2
UN: 3263
ADR: 8 II