Tetrafluoroboric acid ∼50% (w/w) in aqueous solution
Supplier: Thermo Scientific
Danger
Synonyms: Fluoboric acid
L14037.22 L14037.36 L14037.0E
L14037.22EA 28.4 EUR
L14037.22 L14037.36 L14037.0E
Tetrafluoroboric acid ∼50% (w/w) in aqueous solution
Tetrafluoroboric acid
CAS No.: 16872-11-0 Molecular Formula: HBF4 Formula Weight: 87.81 Odour: Pungent Boiling Point: 130° dec. Merck Reference: 14,4149 MDL No.: MFCD00011345
Formula: HBF₄
MW: 87.81 g/mol
Boiling Pt: 130 °C (dec.)
Density: 1.4
Storage Temperature: Ambient
MDL Number: MFCD00011345
CAS Number: 16872-11-0
EINECS: 240-898-3
UN: 1775
ADR: 8,II
Merck Index: 14,4149