Methacrylamide ≥98%
Supplier: Thermo Scientific
Warning
L15013.22 L15013.36
L15013.22EA 26 EUR
L15013.22 L15013.36
Methacrylamide ≥98%
Methacrylamide
Purity: 98% CAS No.: 79-39-0 Molecular Formula: C4H7NO Formula Weight: 85.11 Melting Point: 107-112° Boiling Point: 215° MDL No.: MFCD00008018
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 215 °C
Melting Pt: 107…112 °C
Density: 1.115
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00008018
CAS Number: 79-39-0
EINECS: 201-202-3