Tripotassium orthophosphate ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: Potassium phosphate tribasic, Potassium phosphate
L15168.0P L15168.22 L15168.36
L15168.0PEA 151 EUR
L15168.0P L15168.22 L15168.36
Tripotassium orthophosphate ≥97%
tri-Potassium orthophosphate
Formula: K₃O₄P
MW: 212.27 g/mol
Melting Pt: 1340 °C
Density: 2.564 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036295
CAS Number: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830