1-Butyl-3-methylimidazolium chloride 96%
Supplier: Thermo Scientific
Danger
Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride
L19749.09 L19749.18
L19749.09EA 46.7 EUR
L19749.09 L19749.18
1-Butyl-3-methylimidazolium chloride 96%
1-Butyl-3-methylimidazolium chloride
Purity: 96% CAS No.: 79917-90-1 Molecular Formula: C8H15ClN2 Formula Weight: 174.67 Melting Point: ca 73° MDL No.: MFCD03095425
Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼ 73 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03095425
CAS Number: 79917-90-1
UN: 2811
ADR: 6.1,III